missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NAP1L2 Full-length ORF (AAH26325, 1 a.a. - 460 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH26325 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4674 |
Molecular Weight (g/mol) | 76.34kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16188141
|
Abnova™
H00004674-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178141
|
Abnova™
H00004674-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the nucleosome assembly protein (NAP) family. The function of this family member is unknown; however, mouse studies suggest that it represents a class of tissue-specific factors interacting with chromatin to regulate neuronal cell proliferation. [provided by RefSeq]
Sequence: MAKSENRKELSESSQEEAGNQIMVEGLGEHLERGEDAAAGLGDDGKCGEEAAAGLGEEGENGEDTAAGSGEDGKKGGDTDEDSEADRPKGLIGYVLDTDFVESLPVKVKYRVLALKKLQTRAANLESKFLREFHDIERKFAEMYQPLLEKRRQIINAIYEPTEEECEYKSDSEDCDDEEMCHEEMYGNEEGMVHEYVDEDDGYEDYYYDYAVEEEEEEEEEDDIEATGEENKEEEDPKGIPDFWLTVLKNVDTLTPLIKKYDEPILKLLTDIKVKLSDPGEPLSFTLEFHFKPNEYFKNELLTKTYVLKSKLAYYDPHPYRGTAIEYSTGCEIDWNEGKNVTLKTIKKKQKHRIWGTIRTVTEDFPKDSFFNFFSPHGITSNGRDGNDDFLLGHNLRTYIIPRSVLFFSGDALESQQEGVVREVNDAIYDKIIYDNWMAAIEEVKACCKNLEALVEDIDRSpecifications
AAH26325 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
76.34kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAKSENRKELSESSQEEAGNQIMVEGLGEHLERGEDAAAGLGDDGKCGEEAAAGLGEEGENGEDTAAGSGEDGKKGGDTDEDSEADRPKGLIGYVLDTDFVESLPVKVKYRVLALKKLQTRAANLESKFLREFHDIERKFAEMYQPLLEKRRQIINAIYEPTEEECEYKSDSEDCDDEEMCHEEMYGNEEGMVHEYVDEDDGYEDYYYDYAVEEEEEEEEEDDIEATGEENKEEEDPKGIPDFWLTVLKNVDTLTPLIKKYDEPILKLLTDIKVKLSDPGEPLSFTLEFHFKPNEYFKNELLTKTYVLKSKLAYYDPHPYRGTAIEYSTGCEIDWNEGKNVTLKTIKKKQKHRIWGTIRTVTEDFPKDSFFNFFSPHGITSNGRDGNDDFLLGHNLRTYIIPRSVLFFSGDALESQQEGVVREVNDAIYDKIIYDNWMAAIEEVKACCKNLEALVEDIDR | |
RUO | |
NAP1L2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4674 | |
NAP1L2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BPX/MGC26243 | |
NAP1L2 | |
Recombinant | |
wheat germ expression system |