Learn More
Abnova™ Human NASP Full-length ORF (NP_751896.1, 1 a.a. - 790 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_751896.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4678 |
Molecular Weight (g/mol) | 112.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16180528
|
Abnova™
H00004678-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16190528
|
Abnova™
H00004678-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa. [provided by RefSeq]
Sequence: MFLLLLHLQIKWRATINLLSVTEDGLHFVEYYLNRIIHLDVDSEAKKLLGLGQKHLVMGDIPAAVNAFQEAASLLGKKYGETANECGEAFFFYGKSLLELARMENGVLGNALEGVHVEEEEGEKTEDESLVENNDNIDEEAREELREQVYDAMGEKEEAKKTEDKSLAKPETDKEQDSEMEKGGREDMDISKSAEEPQEKVDLTLDWLTETSEEAKGGAAPEGPNEAEVTSGKPEQEVPDAEEEKSVSGTDVQEECREKGGQEKQGEVIVSIEEKPKEVSEEQPVVTLEKQGTAVEVEAESLDPTVKPVDVGGDEPEEKVVTSENEAGKAVLEQLVGQEVPPAEESPEVTTEAAEASAVEAGSEVSEKPGQEAPVLPKDGAVNGPSVVGDQTPIEPQTSIERLTETKDGSGLEEKVRAKLVPSQEETKLSVEESEAAGDGVDTKVAQGATEKSPEDKVQIAANEETQEREEQMKEGEETEGSEEDDKENDKTEEMPNDSVLENKSLQENEEEEIGNLELAWDMLDLAKIIFKRQETKEAQLYAAQAHLKLGEVSVESENYVQAVEEFQSCLNLQEQYLEAHDRLLAETHYQLGLAYGYNSQYDEAVAQFSKSIEVIENRMAVLNEQVKEAEGSSAEYKKEIEELKELLPEIREKIEDAKESQRSGNVAELALKATLVESSTSGFTPGGGGSSVSMIASRKPTDGASSSNCVTDISHLVRKKRKPEEESPRKDDAKKAKQEPEVNGGSGDAVPSGNEVSENMEEEAENQAESRAAVEGTVEAGATVESTACSpecifications
NP_751896.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
112.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MFLLLLHLQIKWRATINLLSVTEDGLHFVEYYLNRIIHLDVDSEAKKLLGLGQKHLVMGDIPAAVNAFQEAASLLGKKYGETANECGEAFFFYGKSLLELARMENGVLGNALEGVHVEEEEGEKTEDESLVENNDNIDEEAREELREQVYDAMGEKEEAKKTEDKSLAKPETDKEQDSEMEKGGREDMDISKSAEEPQEKVDLTLDWLTETSEEAKGGAAPEGPNEAEVTSGKPEQEVPDAEEEKSVSGTDVQEECREKGGQEKQGEVIVSIEEKPKEVSEEQPVVTLEKQGTAVEVEAESLDPTVKPVDVGGDEPEEKVVTSENEAGKAVLEQLVGQEVPPAEESPEVTTEAAEASAVEAGSEVSEKPGQEAPVLPKDGAVNGPSVVGDQTPIEPQTSIERLTETKDGSGLEEKVRAKLVPSQEETKLSVEESEAAGDGVDTKVAQGATEKSPEDKVQIAANEETQEREEQMKEGEETEGSEEDDKENDKTEEMPNDSVLENKSLQENEEEEIGNLELAWDMLDLAKIIFKRQETKEAQLYAAQAHLKLGEVSVESENYVQAVEEFQSCLNLQEQYLEAHDRLLAETHYQLGLAYGYNSQYDEAVAQFSKSIEVIENRMAVLNEQVKEAEGSSAEYKKEIEELKELLPEIREKIEDAKESQRSGNVAELALKATLVESSTSGFTPGGGGSSVSMIASRKPTDGASSSNCVTDISHLVRKKRKPEEESPRKDDAKKAKQEPEVNGGSGDAVPSGNEVSENMEEEAENQAESRAAVEGTVEAGATVESTAC | |
RUO | |
NASP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4678 | |
NASP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp547F162/FLB7527/FLJ31599/FLJ35510/MGC19722/MGC20372/MGC2297/PRO1999 | |
NASP | |
Recombinant | |
wheat germ expression system |