Learn More
Abnova™ Human NASP Full-length ORF (NP_751896.1, 1 a.a. - 790 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004678-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa. [provided by RefSeq]
Sequence: MFLLLLHLQIKWRATINLLSVTEDGLHFVEYYLNRIIHLDVDSEAKKLLGLGQKHLVMGDIPAAVNAFQEAASLLGKKYGETANECGEAFFFYGKSLLELARMENGVLGNALEGVHVEEEEGEKTEDESLVENNDNIDEEAREELREQVYDAMGEKEEAKKTEDKSLAKPETDKEQDSEMEKGGREDMDISKSAEEPQEKVDLTLDWLTETSEEAKGGAAPEGPNEAEVTSGKPEQEVPDAEEEKSVSGTDVQEECREKGGQEKQGEVIVSIEEKPKEVSEEQPVVTLEKQGTAVEVEAESLDPTVKPVDVGGDEPEEKVVTSENEAGKAVLEQLVGQEVPPAEESPEVTTEAAEASAVEAGSEVSEKPGQEAPVLPKDGAVNGPSVVGDQTPIEPQTSIERLTETKDGSGLEEKVRAKLVPSQEETKLSVEESEAAGDGVDTKVAQGATEKSPEDKVQIAANEETQEREEQMKEGEETEGSEEDDKENDKTEEMPNDSVLENKSLQENEEEEIGNLELAWDMLDLAKIIFKRQETKEAQLYAAQAHLKLGEVSVESENYVQAVEEFQSCLNLQEQYLEAHDRLLAETHYQLGLAYGYNSQYDEAVAQFSKSIEVIENRMAVLNEQVKEAEGSSAEYKKEIEELKELLPEIREKIEDAKESQRSGNVAELALKATLVESSTSGFTPGGGGSSVSMIASRKPTDGASSSNCVTDISHLVRKKRKPEEESPRKDDAKKAKQEPEVNGGSGDAVPSGNEVSENMEEEAENQAESRAAVEGTVEAGATVESTACSpecifications
NP_751896.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
112.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp547F162/FLB7527/FLJ31599/FLJ35510/MGC19722/MGC20372/MGC2297/PRO1999 | |
NASP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4678 | |
NASP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MFLLLLHLQIKWRATINLLSVTEDGLHFVEYYLNRIIHLDVDSEAKKLLGLGQKHLVMGDIPAAVNAFQEAASLLGKKYGETANECGEAFFFYGKSLLELARMENGVLGNALEGVHVEEEEGEKTEDESLVENNDNIDEEAREELREQVYDAMGEKEEAKKTEDKSLAKPETDKEQDSEMEKGGREDMDISKSAEEPQEKVDLTLDWLTETSEEAKGGAAPEGPNEAEVTSGKPEQEVPDAEEEKSVSGTDVQEECREKGGQEKQGEVIVSIEEKPKEVSEEQPVVTLEKQGTAVEVEAESLDPTVKPVDVGGDEPEEKVVTSENEAGKAVLEQLVGQEVPPAEESPEVTTEAAEASAVEAGSEVSEKPGQEAPVLPKDGAVNGPSVVGDQTPIEPQTSIERLTETKDGSGLEEKVRAKLVPSQEETKLSVEESEAAGDGVDTKVAQGATEKSPEDKVQIAANEETQEREEQMKEGEETEGSEEDDKENDKTEEMPNDSVLENKSLQENEEEEIGNLELAWDMLDLAKIIFKRQETKEAQLYAAQAHLKLGEVSVESENYVQAVEEFQSCLNLQEQYLEAHDRLLAETHYQLGLAYGYNSQYDEAVAQFSKSIEVIENRMAVLNEQVKEAEGSSAEYKKEIEELKELLPEIREKIEDAKESQRSGNVAELALKATLVESSTSGFTPGGGGSSVSMIASRKPTDGASSSNCVTDISHLVRKKRKPEEESPRKDDAKKAKQEPEVNGGSGDAVPSGNEVSENMEEEAENQAESRAAVEGTVEAGATVESTAC | |
RUO | |
NASP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |