Learn More
Abnova™ Human NCK2 Partial ORF (NP_003572, 203 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4055.00 SEK - 6150.00 SEK
Specifications
Name | NCK2 (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
For Use With (Application) | Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) |
Molecular Weight | 37.84kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16118545
|
Abnova™
H00008440-Q01.25UG |
25 ug |
6150.00 SEK
25µg |
Estimated Shipment: 11-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
16108545
|
Abnova™
H00008440-Q01.10UG |
10 ug |
4055.00 SEK
10µg |
Estimated Shipment: 11-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Description
This gene encodes a member of the NCK family of adaptor proteins. The protein contains three SH3 domains and one SH2 domain. The protein has no known catalytic function but has been shown to bind and recruit various proteins involved in the regulation of receptor protein tyrosine kinases. It is through these regulatory activities that this protein is believed to be involved in cytoskeletal reorganization. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: LYPFSSVTEEELNFEKGETMEVIEKPENDPEWWKCKNARGQVGLVPKNYVVVLSDGPALHPAHAPQISYTGPSSSGRFAGREWYYGNVTRHQAECALNERGVEGDFLIRDSpecifications
NCK2 (Human) Recombinant Protein (Q01) | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
37.84kDa |