missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NCSTN Partial ORF (AAH47621, 16 a.a. - 115 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023385-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137232

  • 3343.96 SEK / 10µg

Estimated Shipment: 14-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a Type I transmembrane glycoprotein that is an integral component of the multimeric gamma-secretase complex. The encoded protein cleaves integral membrane proteins, including Notch receptors and beta-amyloid precursor protein, and may be a stabilizing cofactor required for gamma-secretase complex assembly. The cleavage of beta-amyloid precursor protein yields amyloid beta peptide, the main component of the neuritic plaque and the hallmark lesion in the brains of patients with Alzheimer's disease; however, the nature of the encoded protein's role in Alzheimer's disease is not known for certain. Alternatively spliced transcript variants have been described, but their full-length nature has not been determined. [provided by RefSeq]

Sequence: VERKIYIPLNKTAPCVRLLNATHQIGCQSSISGDTGVIHVVEKEEDLQWVLTDGPNPPYMVLLESKHFTRDLMEKLKGRTSRIAGLAVSLTKPSPASGFS
Specifications

Specifications

AAH47621
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VERKIYIPLNKTAPCVRLLNATHQIGCQSSISGDTGVIHVVEKEEDLQWVLTDGPNPPYMVLLESKHFTRDLMEKLKGRTSRIAGLAVSLTKPSPASGFS
RUO
NCSTN
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
23385
NCSTN (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
APH2/KIAA0253
NCSTN
Yes
Videos
SDS
Documents
Certificates