missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDUFB5 Full-length ORF (NP_002483.1, 1 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004711-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138221

  • 4055.00 SEK / 10µg
Estimated Shipment: 30-09-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [provided by RefSeq]

Sequence: MAAMSLLRRVSVTAVAALSGRPLGTRLGFGGFLTRGFPKAAAPVRHSGDHGKRLFVIRPSRFYDRRFLKLLRFYIALTGIPVAIFITLVNVFIGQAELAEIPEGYVPEHWEYYKHPISRWIARNFYDSPEKIYERTMAVLQIEAEKAELRVKELEVRKLMHVRGDGPWYYYETIDKELIDHSPKATPDN
Specifications

Specifications

NP_002483.1
Liquid
4711
NDUFB5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAMSLLRRVSVTAVAALSGRPLGTRLGFGGFLTRGFPKAAAPVRHSGDHGKRLFVIRPSRFYDRRFLKLLRFYIALTGIPVAIFITLVNVFIGQAELAEIPEGYVPEHWEYYKHPISRWIARNFYDSPEKIYERTMAVLQIEAEKAELRVKELEVRKLMHVRGDGPWYYYETIDKELIDHSPKATPDN
RUO
NDUFB5
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CI-SGDH/DKFZp686N02262/FLJ30597/MGC111204/MGC12314/SGDH
NDUFB5
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers