Learn More
Abnova™ Human NEB Partial ORF (NP_004534, 2108 a.a. - 2217 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004703-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes nebulin, a giant protein component of the cytoskeletal matrix that coexists with the thick and thin filaments within the sarcomeres of skeletal muscle. In most vertebrates, nebulin accounts for 3 to 4% of the total myofibrillar protein. The encoded protein contains approximately 30-amino acid long modules that can be classified into 7 types and other repeated modules. Protein isoform sizes vary from 600 to 800 kD due to alternative splicing that is tissue-, species-,and developmental stage-specific. Of the 183 exons in the nebulin gene, at least 43 are alternatively spliced, although exons 143 and 144 are not found in the same transcript. Of the several thousand transcript variants predicted for nebulin, the RefSeq Project has decided to create three representative RefSeq records. Mutations in this gene are associated with recessive nemaline myopathy. [provided by RefSeq]
Sequence: NTKTSYHTPADMLSVTAAKDAQANITNTNYKHLIHKYILLPDAMNIELTRNMNRIQSDNEYKQDYNEWYKGLGWSPAGSLEVEKAKKATEYASDQKYRQHPSNFQFKKLTSpecifications
NP_004534 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NTKTSYHTPADMLSVTAAKDAQANITNTNYKHLIHKYILLPDAMNIELTRNMNRIQSDNEYKQDYNEWYKGLGWSPAGSLEVEKAKKATEYASDQKYRQHPSNFQFKKLT | |
RUO | |
NEB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4703 | |
NEB (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686C1456/FLJ11505/FLJ36536/FLJ39568/FLJ39584/NEB177D/NEM2 | |
NEB | |
Recombinant | |
wheat germ expression system |