missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEK3 Partial ORF (AAH19916, 406 a.a. - 506 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH19916 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4752 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16148291
|
Abnova™
H00004752-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138291
|
Abnova™
H00004752-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the NimA (never in mitosis A) family of serine/threonine protein kinases. The encoded protein differs from other NimA family members in that it is not cell cycle regulated and is found primarily in the cytoplasm. The kinase is activated by prolactin stimulation, leading to phosphorylation of VAV2 guanine nucleotide exchange factor, paxillin, and activation of the RAC1 GTPase. Multiple transcript variants encoding different isoforms have been found for this gene
Sequence: QWLKETPDTLLNILKNADLSLAFQTYTIYRPGSEGFLKGPLSEETEASDSVDGGHDSVILDPERLEPGLDEEDTDFEEEDDNPDWVSELKKRAGWQGLCDRSpecifications
AAH19916 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSPK36/MGC29949 | |
NEK3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4752 | |
NEK3 (Human) Recombinant Protein (Q01) | |
QWLKETPDTLLNILKNADLSLAFQTYTIYRPGSEGFLKGPLSEETEASDSVDGGHDSVILDPERLEPGLDEEDTDFEEEDDNPDWVSELKKRAGWQGLCDR | |
RUO | |
NEK3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |