missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEU2 Partial ORF (NP_005374, 180 a.a. - 268 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005374 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4759 |
Molecular Weight (g/mol) | 35.53kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16131845
|
Abnova™
H00004759-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16121845
|
Abnova™
H00004759-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments. [provided by RefSeq]
Sequence: AYRKLHPIQRPIPSAFCFLSHDHGRTWARGHFVAQDTLECQVAEVETGEQRVVTLNARSHLRARVQAQSTNDGLDFQESQLVKKLVEPPSpecifications
NP_005374 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC129579/SIAL2 | |
NEU2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4759 | |
NEU2 (Human) Recombinant Protein (Q01) | |
AYRKLHPIQRPIPSAFCFLSHDHGRTWARGHFVAQDTLECQVAEVETGEQRVVTLNARSHLRARVQAQSTNDGLDFQESQLVKKLVEPP | |
RUO | |
NEU2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |