missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEUROD2 Partial ORF (NP_006151.2, 266 a.a. - 375 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006151.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4761 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16181845
|
Abnova™
H00004761-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16171845
|
Abnova™
H00004761-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the neuroD family of neurogenic basic helix-loop-helix (bHLH) proteins. Expression of this gene can induce transcription from neuron-specific promoters, such as the GAP-43 promoter, which contain a specific DNA sequence known as an E-box. The product of the human gene can induce neurogenic differentiation in non-neuronal cells in Xenopus embryos, and is thought to play a role in the determination and maintenance of neuronal cell fates. [provided by RefSeq]
Sequence: RTHGYCAAYETLYAAAGGGGASPDYNSSEYEGPLSPPLCLNGNFSLKQDSSPDHEKSYHYSMHYSALPGSRPTGHGLVFGSSAVRGGVHSENLLSYDMHLHHDRGPMYEESpecifications
NP_006151.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC26304/NDRF/bHLHa1 | |
NEUROD2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4761 | |
NEUROD2 (Human) Recombinant Protein (Q01) | |
RTHGYCAAYETLYAAAGGGGASPDYNSSEYEGPLSPPLCLNGNFSLKQDSSPDHEKSYHYSMHYSALPGSRPTGHGLVFGSSAVRGGVHSENLLSYDMHLHHDRGPMYEE | |
RUO | |
NEUROD2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |