missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NEUROD6 Full-length ORF (AAH35048, 1 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
3970.00 SEK - 6025.00 SEK
Specifications
Accession Number | AAH35048 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 63974 |
Molecular Weight (g/mol) | 62.81kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16183333
|
Abnova™
H00063974-P01.10UG |
10 ug |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16193333
|
Abnova™
H00063974-P01.25UG |
25 ug |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
NEUROD6 is a member of the NEUROD (NEUROD1; MIM 601724) family of basic helix-loop-helix (bHLH) transcription factors (Guo et al., 2002 [PubMed 12357074]).[supplied by OMIM]
Sequence: MLTLPFDESVVMPESQMCRKFSRECEDQKQIKKPESFSKQIVLRGKSIKRAPGEETEKEEEEEDREEEDENGLPRRRGLRKKKTTKLRLERVKFRRQEANARERNRMHGLNDALDNLRKVVPCYSKTQKLSKIETLRLAKNYIWALSEILRIGKRPDLLTFVQNLCKGLSQPTTNLVAGCLQLNARSFMMGQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMHYCAVPPRGPLGQGAMFRLPTDSHFPYDLHLRSQSLTMQDELNAVFHNSpecifications
AAH35048 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.81kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLTLPFDESVVMPESQMCRKFSRECEDQKQIKKPESFSKQIVLRGKSIKRAPGEETEKEEEEEDREEEDENGLPRRRGLRKKKTTKLRLERVKFRRQEANARERNRMHGLNDALDNLRKVVPCYSKTQKLSKIETLRLAKNYIWALSEILRIGKRPDLLTFVQNLCKGLSQPTTNLVAGCLQLNARSFMMGQGGEAAHHTRSPYSTFYPPYHSPELTTPPGHGTLDNSKSMKPYNYCSAYESFYESTSPECASPQFEGPLSPPPINYNGIFSLKQEETLDYGKNYNYGMHYCAVPPRGPLGQGAMFRLPTDSHFPYDLHLRSQSLTMQDELNAVFHN | |
RUO | |
NEUROD6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
63974 | |
NEUROD6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Atoh2/MATH2/Math-2/NEX1M/bHLHa2 | |
NEUROD6 | |
Recombinant | |
wheat germ expression system |