Läs mer
Human Neuroligin 4X/NLGN4X Antibody, R&D Systems™
Produktkod. 18023322
Change view
Kvantitet:
100 μg
Produktkod. Kvantitet
18023322 100 μg
1 options
Produktkod. 18023322 Leverantör R&D Systems Leverantörsnummer AF5158
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Sheep Polyclonal Antibody
Produced in sheep immunized with purified, E. coli-derived, recombinant human Neuroligin 4 extracellular domain (rhNLGN4; R&D Systems, Catalog # 5158-NL). Human NLGN4 specific IgG was purified by human NLGN4 affinity chromatography.
Neuroligin 4X/NLGN4X Polyclonal specifically detects Neuroligin 4X/NLGN4X in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifikationer
Antigen Neuroligin 4X/NLGN4X
Användningsområden Western Blot, Immunohistochemistry
Klassificering Polyclonal
Koncentration LYOPH
Konjugera Unconjugated
Utspädning Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Genaccessionsnr. Q8N0W4
Gene Alias ASPGX2, AUTSX2, HLNX, HNL4X, HNLX, KIAA1260AUTSX2, MGC22376, neuroligin 4, X-linked, NL4, NLGN, NLGN4, NLGN4neuroligin 4, NLGN4X, X-linked
Gensymboler NLGN4X
Värd art Sheep
Immunogen E. coli-derived recombinant human Neuroligin 4/NLGN4 Met618-Thr673 Accession # Q8N0W4
Reningsmetod Affinity Purified
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 57502
Testspecificitet Detects human Neuroligin 4/NLGN4 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) NLGN3 is observed.
Rekonstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Målarter Human
Innehåll och lagring Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotyp IgG