Learn More
Abnova™ Human NFAM1 Partial ORF (NP_666017.1, 187 a.a. - 269 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00150372-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a type I membrane receptor that activates cytokine gene promoters such as the IL-13 and TNF-alpha promoters. The encoded protein contains an immunoreceptor tyrosine-based activation motif (ITAM) and is thought to regulate the signaling and development of B-cells. [provided by RefSeq]
Sequence: NKKRMRGPGKDPTRKCPDPRSASSPKQHPSESVYTALQRRETEVYACIENEDGSSPTAKQSPLSQERPHRFEDDGELNLVYENSpecifications
NP_666017.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.87kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NKKRMRGPGKDPTRKCPDPRSASSPKQHPSESVYTALQRRETEVYACIENEDGSSPTAKQSPLSQERPHRFEDDGELNLVYEN | |
RUO | |
NFAM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
150372 | |
NFAM1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CNAIP/FLJ40652/bK126B4.4 | |
NFAM1 | |
Recombinant | |
wheat germ expression system |