missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NFKBIB Full-length ORF (NP_002494.2, 1 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004793-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180718

  • 3343.96 SEK / 10µg

Estimated Shipment: 09-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL (MIM 164910), RELA (MIM 164014), or RELB (MIM 604758) to form the NFKB complex. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA, MIM 164008, or NFKBIB), which inactivate NF-kappa-B by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664 or IKBKB, MIM 603258) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NF-kappa-B complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine).[supplied by OMIM]

Sequence: MAGVACLGKAADADEWCDSGLGSLGPDAAAPGGPGLGAELGPGLSWAPLVFGYVTEDGDTALHLAVIHQHEPFLDFLLGFSAGTEYMDLQNDLGQTALHLAAILGETSTVEKLYAAGAGLCVAERRGHTALHLACRVGAHACARALLQPRPRRPREAPDTYLAQGPDRTPDTNHTPVALYPDSDLEKEEEESEEDWKLQLEAENYEGHTPLHVAVIHKDVEMVRLLRDAGADLDKPEPTCGRSPLHLAVEAQAADVLELLLRAGANPAARMYGGRTPLGSAMLRPNPILARLLRAHGAPEPEGEDEKSGPCSSSSDSDSGDEGDEYDDIVVHSSRSQTRLPPTPASKPLPDDPRPV
Specifications

Specifications

NP_002494.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
64.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IKBB/TRIP9
NFKBIB
Yes
wheat germ expression system
Liquid
4793
NFKBIB (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGVACLGKAADADEWCDSGLGSLGPDAAAPGGPGLGAELGPGLSWAPLVFGYVTEDGDTALHLAVIHQHEPFLDFLLGFSAGTEYMDLQNDLGQTALHLAAILGETSTVEKLYAAGAGLCVAERRGHTALHLACRVGAHACARALLQPRPRRPREAPDTYLAQGPDRTPDTNHTPVALYPDSDLEKEEEESEEDWKLQLEAENYEGHTPLHVAVIHKDVEMVRLLRDAGADLDKPEPTCGRSPLHLAVEAQAADVLELLLRAGANPAARMYGGRTPLGSAMLRPNPILARLLRAHGAPEPEGEDEKSGPCSSSSDSDSGDEGDEYDDIVVHSSRSQTRLPPTPASKPLPDDPRPV
RUO
NFKBIB
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates