Learn More
Abnova™ Human NHLRC1 Partial ORF (NP_940988.2, 137 a.a. - 246 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00378884-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The NHLRC1 gene encodes malin, a single subunit E3 ubiquitin (UBB; MIM 191339) ligase, which contains a RING-HC-type zinc finger and 6 NHL domains and is subclassified as a member of the RING-HCa family (Gentry et al., 2005 [PubMed 15930137]).[supplied by OMIM]
Sequence: KTGRVVVVHDGRRRVKIFDSGGGCAHQFGEKGDAAQDIRYPVDVTITNDCHVVVTDAGDRSIKVFDFFGQIKLVIGGQFSLPWGVETTPQNGIVVTDAEAGSLHLLDVDFSpecifications
NP_940988.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KTGRVVVVHDGRRRVKIFDSGGGCAHQFGEKGDAAQDIRYPVDVTITNDCHVVVTDAGDRSIKVFDFFGQIKLVIGGQFSLPWGVETTPQNGIVVTDAEAGSLHLLDVDF | |
RUO | |
NHLRC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
378884 | |
NHLRC1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EPM2A/EPM2B/MALIN/MGC119262/MGC119264/MGC119265/bA204B7.2 | |
NHLRC1 | |
Recombinant | |
wheat germ expression system |