missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NIBP Full-length ORF (AAH06206.3, 1 a.a. - 912 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH06206.3 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 83696 |
Molecular Weight (g/mol) | 127.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16135433
|
Abnova™
H00083696-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16125433
|
Abnova™
H00083696-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NIBP binds NIK (MIM 604655) and IKK-beta (IKBKB; MIM 603258) and plays a role in the neuronal NF-kappa-B (see MIM 164011) signaling pathway (Hu et al., 2005 [PubMed 15951441]).[supplied by OMIM]
Sequence: MSVELLRSVNDFLWLGAALEGLCSASVIYHYPGGTGGKSGARRFQGSTLPAEAANRHRPGALTTNGINPDTSTEIGRAKNCLSPEDIIDKYKEAISYYSKYKNAGVIELEACIKAVRVLAIQKRSMEASEFLQNAVYINLRQLSEEEKIQRYSILSELYELIGFHRKSAFFKRVAAMQCVAPSIAEPGWRACYKLLLETLPGYSLSLDPKDFSRGTHRGWAAVQMRLLHELVYASRRMGNPALSVRHLSFLLQTMLDFLSDQEKKDVAQSLENYTSKCPGTMEPIALPGGLTLPPVPFTKLPIVRHVKLLNLPASLRPHKMKSLLGQNVSTKSPFIYSPIIAHNRGEERNKKIDFQWVQGDVCEVQLMVYNPMPFELRVENMGLLTSGVEFESLPAALSLPAESGLYPVTLVGVPQTTGTITVNGYHTTVFGVFSDCLLDNLPGIKTSGSTVEVIPALPRLQISTSLPRSAHSLQPSSGDEISTNVSVQLYNGESQQLIIKLENIGMEPLEKLEVTSKVLTTKEKLYGDFLSWKLEETLAQFPLQPGKVATFTINIKVKLDFSCQENLLQDLSDDGISVSGFPLSSPFRQVVRPRVEGKPVNPPESNKAGDYSHVKTLEAVLNFKYSGGPGHTEGYYRNLSLGLHVEVEPSVFFTRVSTLPATSTRQCHLLLDVFNSTEHELTVSTRSSEALILHAGECQRMAIQVDKFNFESFPESPGEKGQFANPKQLEEERREARGLEIHSKLGICWRIPSLKRSGEASVEGLLNQLVLEHLQLAPLQWDVLVDGQPCDREAVAACQVGDPVRLEVRLTNRSPRSVGPFALTVVPFQDHQNGVHNYDLHDTVSFVGSSTFYLDAVQPSGQSACLGALLFLYTGDFFLHIRFHEDSTSKELPPSWFCLPSVHVCALEAQASpecifications
AAH06206.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
127.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSVELLRSVNDFLWLGAALEGLCSASVIYHYPGGTGGKSGARRFQGSTLPAEAANRHRPGALTTNGINPDTSTEIGRAKNCLSPEDIIDKYKEAISYYSKYKNAGVIELEACIKAVRVLAIQKRSMEASEFLQNAVYINLRQLSEEEKIQRYSILSELYELIGFHRKSAFFKRVAAMQCVAPSIAEPGWRACYKLLLETLPGYSLSLDPKDFSRGTHRGWAAVQMRLLHELVYASRRMGNPALSVRHLSFLLQTMLDFLSDQEKKDVAQSLENYTSKCPGTMEPIALPGGLTLPPVPFTKLPIVRHVKLLNLPASLRPHKMKSLLGQNVSTKSPFIYSPIIAHNRGEERNKKIDFQWVQGDVCEVQLMVYNPMPFELRVENMGLLTSGVEFESLPAALSLPAESGLYPVTLVGVPQTTGTITVNGYHTTVFGVFSDCLLDNLPGIKTSGSTVEVIPALPRLQISTSLPRSAHSLQPSSGDEISTNVSVQLYNGESQQLIIKLENIGMEPLEKLEVTSKVLTTKEKLYGDFLSWKLEETLAQFPLQPGKVATFTINIKVKLDFSCQENLLQDLSDDGISVSGFPLSSPFRQVVRPRVEGKPVNPPESNKAGDYSHVKTLEAVLNFKYSGGPGHTEGYYRNLSLGLHVEVEPSVFFTRVSTLPATSTRQCHLLLDVFNSTEHELTVSTRSSEALILHAGECQRMAIQVDKFNFESFPESPGEKGQFANPKQLEEERREARGLEIHSKLGICWRIPSLKRSGEASVEGLLNQLVLEHLQLAPLQWDVLVDGQPCDREAVAACQVGDPVRLEVRLTNRSPRSVGPFALTVVPFQDHQNGVHNYDLHDTVSFVGSSTFYLDAVQPSGQSACLGALLFLYTGDFFLHIRFHEDSTSKELPPSWFCLPSVHVCALEAQA | |
RUO | |
TRAPPC9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
83696 | |
NIBP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IBP/IKBKBBP/KIAA1882/MGC4737/MGC4769/NIBP/T1/TRS120 | |
TRAPPC9 | |
Recombinant | |
wheat germ expression system |