Learn More
Abnova™ Human NLGN1 Partial ORF (NP_055747, 578 a.a. - 677 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00022871-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. [provided by RefSeq]
Sequence: YSQKDQLYLHIGLKPRVKEHYRANKVNLWLELVPHLHNLNDISQYTSTTTKVPSTDITFRPTRKNSVPVTSAFPTAKQDDPKQQPSPFSVDQRDYSTELSSpecifications
NP_055747 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YSQKDQLYLHIGLKPRVKEHYRANKVNLWLELVPHLHNLNDISQYTSTTTKVPSTDITFRPTRKNSVPVTSAFPTAKQDDPKQQPSPFSVDQRDYSTELS | |
RUO | |
NLGN1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
22871 | |
NLGN1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1070/MGC45115 | |
NLGN1 | |
Recombinant | |
wheat germ expression system |