missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NLGN3 Full-length ORF (AAH51715.1, 1 a.a. - 828 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH51715.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 54413 |
Molecular Weight (g/mol) | 118.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16120523
|
Abnova™
H00054413-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16110523
|
Abnova™
H00054413-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene, but their full length sequences have not been determined. [provided by RefSeq]
Sequence: MWLRLGPPSLSLSPKPTVGRSLCLTLWFLSLALRASTQAPAPTVNTHFGKLRGARVPLPSEILGPVDQYLGVPYAAPPIGEKRFLPPEPPPYWSGIRNATHFPPVCPQNIHTAVPEVMLPVWFTANLDIVATYIQEPNEDCLYLNVYVPTEDGSGAKKQGEDLADNDGDEDEDIRDSGAKPVMVYIHGGSYMEGTGNMIDGSILASYGNVIVITLNYRVGVLGFLSTGDQAAKGNYGLLDQIQALRWVSENIAFFGGDPRRITVFGSGIGASCVSLLTLSHHSEGLFQRAIIQSGSALSSWAVNYQPVKYTSLLADKVGCNVLDTVDMVDCLRQKSAKELVEQDIQPARYHVAFGPVIDGDVIPDDPEILMEQGEFLNYDIMLGVNQGEGLKFVEGVVDPEDGVSGTDFDYSVSNFVDNLYGYPEGKDTLRETIKFMYTDWADRDNPETRRKTLVALFTDHQWVEPSVVTADLHARYGSPTYFYAFYHHCQSLMKPAWSDAAHGDEVPYVFGVPMVGPTDLFPCNFSKNDVMLSAVVMTYWTNFAKTGDPNKPVPQDTKFIHTKANRFEEVAWSKYNPRDQLYLHIGLKPRVRDHYRATKVAFWKHLVPHLYNLHDMFHYTSTTTKVPPPDTTHSSHITRRPNGKTWSTKRPAISPAYSNENAQGSWNGDQDAGPLLVENPRDYSTELSVTIAVGASILFLNVLAFAALYYRKDKRRQEPLRQPSPQRGAWAPELGAAPEEELAALQLGPTHHECEASPPHDTLRLTALPDYTLTLRRSPDDIPLMTPNTITMIPNSLVGLQTLHPYNTFAAGFNSTGLPHSHSTTRVSpecifications
AAH51715.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MWLRLGPPSLSLSPKPTVGRSLCLTLWFLSLALRASTQAPAPTVNTHFGKLRGARVPLPSEILGPVDQYLGVPYAAPPIGEKRFLPPEPPPYWSGIRNATHFPPVCPQNIHTAVPEVMLPVWFTANLDIVATYIQEPNEDCLYLNVYVPTEDGSGAKKQGEDLADNDGDEDEDIRDSGAKPVMVYIHGGSYMEGTGNMIDGSILASYGNVIVITLNYRVGVLGFLSTGDQAAKGNYGLLDQIQALRWVSENIAFFGGDPRRITVFGSGIGASCVSLLTLSHHSEGLFQRAIIQSGSALSSWAVNYQPVKYTSLLADKVGCNVLDTVDMVDCLRQKSAKELVEQDIQPARYHVAFGPVIDGDVIPDDPEILMEQGEFLNYDIMLGVNQGEGLKFVEGVVDPEDGVSGTDFDYSVSNFVDNLYGYPEGKDTLRETIKFMYTDWADRDNPETRRKTLVALFTDHQWVEPSVVTADLHARYGSPTYFYAFYHHCQSLMKPAWSDAAHGDEVPYVFGVPMVGPTDLFPCNFSKNDVMLSAVVMTYWTNFAKTGDPNKPVPQDTKFIHTKANRFEEVAWSKYNPRDQLYLHIGLKPRVRDHYRATKVAFWKHLVPHLYNLHDMFHYTSTTTKVPPPDTTHSSHITRRPNGKTWSTKRPAISPAYSNENAQGSWNGDQDAGPLLVENPRDYSTELSVTIAVGASILFLNVLAFAALYYRKDKRRQEPLRQPSPQRGAWAPELGAAPEEELAALQLGPTHHECEASPPHDTLRLTALPDYTLTLRRSPDDIPLMTPNTITMIPNSLVGLQTLHPYNTFAAGFNSTGLPHSHSTTRV | |
RUO | |
NLGN3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
54413 | |
NLGN3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ASPGX1/AUTSX1/HNL3/KIAA1480 | |
NLGN3 | |
Recombinant | |
wheat germ expression system |