missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NNAT Full-length ORF (AAH01768, 1 a.a. - 81 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004826-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16178391

  • 3343.96 SEK / 10µg

Estimated Shipment: 09-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a proteolipid that may be involved in the regulation of ion channels during brain development. The encoded protein may also play a role in forming and maintaining the structure of the nervous system. This gene is found within an intron of the BLCAP gene, but on the opposite strand. This gene is imprinted and is expressed only from the paternal allele, while BLCAP is not imprinted. Two transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MAAVAAASAELLIIGWYIFRVLLQVFLECCIYWVGFAFRNPPGTQPIARSEVFRYSLQKLAYTVSRTGRQVLGERRQRAPN
Specifications

Specifications

AAH01768
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.65kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC1439/Peg5
NNAT
Yes
wheat germ expression system
Liquid
4826
NNAT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAVAAASAELLIIGWYIFRVLLQVFLECCIYWVGFAFRNPPGTQPIARSEVFRYSLQKLAYTVSRTGRQVLGERRQRAPN
RUO
NNAT
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates