Learn More
Abnova™ Human NOMO3 Partial ORF (NP_001004067.1, 966 a.a. - 1033 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00408050-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein originally thought to be related to the collagenase gene family. This gene is one of three highly similar genes in a duplicated region on the short arm of chromosome 16. These three genes encode closely related proteins that may have the same function. The protein encoded by one of these genes has been identified as part of a protein complex that participates in the Nodal signaling pathway during vertebrate development. Mutations in ABCC6, which is located nearby, rather than mutations in this gene are associated with pseudoxanthoma elasticum. [provided by RefSeq]
Sequence: TVSSLNGEPEQGVAMEAVGQNDCSIYGEDTVTDEEGKFRLRGLLPGCVYHVQLKAEGNDHIERALPHHSpecifications
NP_001004067.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.22kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TVSSLNGEPEQGVAMEAVGQNDCSIYGEDTVTDEEGKFRLRGLLPGCVYHVQLKAEGNDHIERALPHH | |
RUO | |
NOMO3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
408050 | |
NOMO3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Nomo | |
NOMO3 | |
Recombinant | |
wheat germ expression system |