Learn More
Abnova™ Human NOS1AP Full-length ORF (NP_055512.1, 1 a.a. - 506 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009722-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a cytosolic protein that binds to the signaling molecule, neuronal nitric oxide synthase (nNOS). This protein has a C-terminal PDZ-binding domain that mediates interactions with nNOS and an N-terminal phosphotyrosine binding (PTB) domain that binds to the small monomeric G protein, Dexras1. Studies of the related mouse and rat proteins have shown that this protein functions as an adapter protein linking nNOS to specific targets, such as Dexras1 and the synapsins. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MPSKTKYNLVDDGHDLRIPLHNEDAFQHGICFEAKYVGSLDVPRPNSRVEIVAAMRRIRYEFKAKNIKKKKVSIMVSVDGVKVILKKKKKLLLLQKKEWTWDESKMLVMQDPIYRIFYVSHDSQDLKIFSYIARDGASNIFRCNVFKSKKKSQAMRIVRTVGQAFEVCHKLSLQHTQQNADGQEDGESERNSNSSGDPGRQLTGAERASTATAEETDIDAVEVPLPGNDVLEFSRGVTDLDAVGKEGGSHTGSKVSHPQEPMLTASPRMLLPSSSSKPPGLGTETPLSTHHQMQLLQQLLQQQQQQTQVAVAQVHLLKDQLAAEAAARLEAQARVHQLLLQNKDMLQHISLLVKQVQELELKLSGQNAMGSQDSLLEITFRSGALPVLCDPTTPKPEDLHSPPLGAGLADFAHPAGSPLGRRDCLVKLECFRFLPPEDTPPPAQGEALLGGLELIKFRESGIASEYESNTDESEERDSWSQEELPRLLNVLQRQELGDGLDDEIAVSpecifications
NP_055512.1 | |
Liquid | |
9722 | |
NOS1AP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPSKTKYNLVDDGHDLRIPLHNEDAFQHGICFEAKYVGSLDVPRPNSRVEIVAAMRRIRYEFKAKNIKKKKVSIMVSVDGVKVILKKKKKLLLLQKKEWTWDESKMLVMQDPIYRIFYVSHDSQDLKIFSYIARDGASNIFRCNVFKSKKKSQAMRIVRTVGQAFEVCHKLSLQHTQQNADGQEDGESERNSNSSGDPGRQLTGAERASTATAEETDIDAVEVPLPGNDVLEFSRGVTDLDAVGKEGGSHTGSKVSHPQEPMLTASPRMLLPSSSSKPPGLGTETPLSTHHQMQLLQQLLQQQQQQTQVAVAQVHLLKDQLAAEAAARLEAQARVHQLLLQNKDMLQHISLLVKQVQELELKLSGQNAMGSQDSLLEITFRSGALPVLCDPTTPKPEDLHSPPLGAGLADFAHPAGSPLGRRDCLVKLECFRFLPPEDTPPPAQGEALLGGLELIKFRESGIASEYESNTDESEERDSWSQEELPRLLNVLQRQELGDGLDDEIAV | |
RUO | |
NOS1AP | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
6330408P19Rik/CAPON/MGC138500 | |
NOS1AP | |
Yes | |
wheat germ expression system |