Learn More
Abnova™ Human NOVA1 Partial ORF (NP_002506, 408 a.a. - 507 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004857-Q01.25ug
Additional Details : Weight : 0.02000kg
Description
This gene encodes a neuron-specific RNA-binding protein, a member of the Nova family of paraneoplastic disease antigens, that is recognized and inhibited by paraneoplastic antibodies. These antibodies are found in the sera of patients with paraneoplastic opsoclonus-ataxia, breast cancer, and small cell lung cancer. Alternatively spliced transcripts encoding distinct isoforms have been described. [provided by RefSeq]
Sequence: SAILGTEKSTDGSKDVVEIAVPENLVGAILGKGGKTLVEYQELTGARIQISKKGEFVPGTRNRKVTITGTPAATQAAQYLITQRITYEQGVRAANPQKVGSpecifications
NP_002506 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SAILGTEKSTDGSKDVVEIAVPENLVGAILGKGGKTLVEYQELTGARIQISKKGEFVPGTRNRKVTITGTPAATQAAQYLITQRITYEQGVRAANPQKVG | |
RUO | |
NOVA1 | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
4857 | |
NOVA1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Nova-1 | |
NOVA1 | |
Yes |