Learn More
Abnova™ Human NPAS2 Full-length ORF (AAH51351.2, 1 a.a. - 824 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004862-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. A similar mouse protein may play a regulatory role in the acquisition of specific types of memory. It also may function as a part of a molecular clock operative in the mammalian forebrain. [provided by RefSeq]
Sequence: MDEDEKDRAKRASRNKSEKKRRDQFNVLIKELSSMLPGNTRKMDKTTVLEKVIGFLQKHNEVSAQTEICDIQQDWKPSFLSNEEFTQLMLEALDGFIIAVTTDGSIIYVSDSITPLLGHLPSDVMDQNLLNFLPEQEHSEVYKILSSHMLVTDSPSPEYLKSDSDLEFYCHLLRGSLNPKEFPTYEYIKFVGNFRSYNNVPSPSCNGFDNTLSRPCRVPLGKEVCFIATVRLATPQFLKEMCIVDEPLEEFTSRHSLEWKFLFLDHRAPPIIGYLPFEVLGTSGYDYYHIDDLELLARCHQHLMQFGKGKSCCYRFLTKGQQWIWLQTHYYITYHQWNSKPEFIVCTHSVVSYADVRVERRQELALEDPPSEALHSSALKDKGSSLEPRQHFNALDVGASGLNTSHSPSASSRSSHKSSHTAMSEPTSTPTKLMAEASTPALPRSATLPQELPVPGLSQAATMPAPLPSPSSCDLTQQLLPQTVLQSTPAPMAQFSAQFSMFQTIKDQLEQRTRILQANIRWQQEELHKIQEQLCLVQDSNVQMFLQQPAVSLSFSSTQRPEAQQQLQQRSAAVTQPQLGAGPQLPGQISSAQVTSQHLLRESSVISTQGPKPMRSSQLMQSSGRSGSSLVSPFSSATAALPPSLNLTTPASTSQDASQCQPSPDFSHDRQLRLLLSQPIQPMMPGSCDARQPSEVSRTGRQVKYAQSQTVFQNPDAHPANSSSAPMPVLLMGQAVLHPSFPASQPSPLQPAQARQQPPQHYLQVQAPTSLHSEQQDSLLLSTYSQQPGTLGYPQPPPAQPQPLRPPRRVSSLSESSGLQQPPRSpecifications
AAH51351.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23138/MGC71151/MOP4/PASD4/bHLHe9 | |
NPAS2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4862 | |
NPAS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDEDEKDRAKRASRNKSEKKRRDQFNVLIKELSSMLPGNTRKMDKTTVLEKVIGFLQKHNEVSAQTEICDIQQDWKPSFLSNEEFTQLMLEALDGFIIAVTTDGSIIYVSDSITPLLGHLPSDVMDQNLLNFLPEQEHSEVYKILSSHMLVTDSPSPEYLKSDSDLEFYCHLLRGSLNPKEFPTYEYIKFVGNFRSYNNVPSPSCNGFDNTLSRPCRVPLGKEVCFIATVRLATPQFLKEMCIVDEPLEEFTSRHSLEWKFLFLDHRAPPIIGYLPFEVLGTSGYDYYHIDDLELLARCHQHLMQFGKGKSCCYRFLTKGQQWIWLQTHYYITYHQWNSKPEFIVCTHSVVSYADVRVERRQELALEDPPSEALHSSALKDKGSSLEPRQHFNALDVGASGLNTSHSPSASSRSSHKSSHTAMSEPTSTPTKLMAEASTPALPRSATLPQELPVPGLSQAATMPAPLPSPSSCDLTQQLLPQTVLQSTPAPMAQFSAQFSMFQTIKDQLEQRTRILQANIRWQQEELHKIQEQLCLVQDSNVQMFLQQPAVSLSFSSTQRPEAQQQLQQRSAAVTQPQLGAGPQLPGQISSAQVTSQHLLRESSVISTQGPKPMRSSQLMQSSGRSGSSLVSPFSSATAALPPSLNLTTPASTSQDASQCQPSPDFSHDRQLRLLLSQPIQPMMPGSCDARQPSEVSRTGRQVKYAQSQTVFQNPDAHPANSSSAPMPVLLMGQAVLHPSFPASQPSPLQPAQARQQPPQHYLQVQAPTSLHSEQQDSLLLSTYSQQPGTLGYPQPPPAQPQPLRPPRRVSSLSESSGLQQPPR | |
RUO | |
NPAS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |