Läs mer
Human NPDC-1 Antibody, R&D Systems™
GREENER_CHOICE
Produktkod. 18032112
Change view
Kvantitet:
100 μg
Produktkod. Kvantitet
18032112 100 μg
1 options
Produktkod. 18032112 Leverantör R&D Systems Leverantörsnummer AF5489
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Sheep Polyclonal Antibody
NPDC-1 Polyclonal specifically detects NPDC-1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifikationer
Antigen NPDC-1
Användningsområden Western Blot, Immunohistochemistry
Klassificering Polyclonal
Koncentration LYOPH
Konjugera Unconjugated
Utspädning Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Genaccessionsnr. Q9NQX5
Gene Alias CAB-, CAB1, CAB-1, CAB1CAB, DKFZp586J0523, neural proliferation differentiation and control protein 1, neural proliferation, differentiation and control, 1, NPDC1, NPDC-1
Gensymboler NPDC1
Värd art Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human NPDC-1 Gly35-Asp181 Accession # Q9NQX5
Reningsmetod Affinity Purified
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 56654
Testspecificitet Detects human NPDC-1 in direct ELISAs and Western blots.
Rekonstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Målarter Human
Innehåll och lagring Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotyp IgG