Learn More
Abnova™ Human NPHP4 Full-length ORF (AAH50076.1, 1 a.a. - 81 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00261734-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein which contains a proline-rich region. The encoded protein may function in renal tubular development and function. This protein interacts with nephrocystin. Mutations in this gene are associated with nephronophthisis type 4. Multiple alternative transcript variants have been described but their full-length nature has not been determined. [provided by RefSeq]
Sequence: MCGAFSTQAGAVEKTLFDISPAPGAPVGMLGEDPPVHVRCSDPNVICETQNVGPGEPRDIFLKVASGPSPEIKDFFVIIYSSpecifications
AAH50076.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0673/SLSN4 | |
NPHP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
261734 | |
NPHP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MCGAFSTQAGAVEKTLFDISPAPGAPVGMLGEDPPVHVRCSDPNVICETQNVGPGEPRDIFLKVASGPSPEIKDFFVIIYS | |
RUO | |
NPHP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |