missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NR2F2 Partial ORF (NP_066285, 283 a.a. - 342 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007026-Q02.10ug
Additional Details : Weight : 0.02000kg
Description
Sequence: DRVVAFMDHIRIFQEQVEKLKALHVDSAEYSCLKAIVLFTSDACGLSDVAHVESLQEKSQSpecifications
NP_066285 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.34kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
DRVVAFMDHIRIFQEQVEKLKALHVDSAEYSCLKAIVLFTSDACGLSDVAHVESLQEKSQ | |
RUO | |
NR2F2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7026 | |
NR2F2 (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARP1/COUP-TFII/COUPTFB/MGC117452/SVP40/TFCOUP2 | |
NR2F2 | |
Recombinant | |
wheat germ expression system |