Learn More
Abnova™ Human NSBP1 Full-length ORF (NP_110390.1, 1 a.a. - 282 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079366-P02.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a nuclear protein with similarities to the high mobility group proteins, HMG14 and HMG17, which suggests that this protein may function as a nucleosomal binding and transcriptional activating protein. [provided by RefSeq]
Sequence: MPKRKAAGQGDMRQEPKRRSARLSAMLVPVTPEVKPKRTSSSRKMKTKSDMMEENIDTSAQAVAETKQEAVVEEDYNENAKNGEAKITEAPASEKEIVEVKEENIEDATEKGGEKKEAVAAEVKNEEEDQKEDEEDQNEEKGEAGKEDKDEKGEEDGKEDKNGNEKGEDAKEKEDGKKGEDGKGNGEDGKEKGEDEKEEEDRKETGDGKENEDGKEKGDKKEGKDVKVKEDEKEREDGKEDEGGNEEEAGKEKEDLKEEEEGKEEDEIKEDDGKKEEPQSIVSpecifications
NP_110390.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NSBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
79366 | |
NSBP1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPKRKAAGQGDMRQEPKRRSARLSAMLVPVTPEVKPKRTSSSRKMKTKSDMMEENIDTSAQAVAETKQEAVVEEDYNENAKNGEAKITEAPASEKEIVEVKEENIEDATEKGGEKKEAVAAEVKNEEEDQKEDEEDQNEEKGEAGKEDKDEKGEEDGKEDKNGNEKGEDAKEKEDGKKGEDGKGNGEDGKEKGEDEKEEEDRKETGDGKENEDGKEKGDKKEGKDVKVKEDEKEREDGKEDEGGNEEEAGKEKEDLKEEEEGKEEDEIKEDDGKKEEPQSIV | |
RUO | |
NSBP1 | |
Recombinant | |
wheat germ expression system |