Läs mer
Gibco™ Human NT-3 Recombinant Protein
Beskrivning
Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute NT-3 in water, to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.
Specifikationer
Specifikationer
| Tillträdesnummer | P20783 |
| För användning med (applikation) | Bioactivity |
| Formulering | Protein with no preservative |
| Gen-ID (Entrez) | 4908 |
| Namn | Human NT-3 |
| Reningsmetod | Purified |
| Kvantitet | 10 μg |
| Förvaringskrav | -20°C |
| Regulatorisk status | RUO |
| Endotoxinkoncentration | <0.1 ng/μg |
| Visa mer |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.