Läs mer
Abnova™ Human NTRK1 (NM_002529.2, 443 a.a. - 796 a.a.) Partial Recombinant Protein with GST-His tag expressed in Sf9 cells
Beskrivning
This gene encodes a member of the neurotrophic tyrosine kinase receptor (NTKR) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of this kinase leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in this gene have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer. Alternate transcriptional splice variants of this gene have been found, but only three have been characterized to date. [provided by RefSeq]
Specifikationer
Specifikationer
| Tillträdesnummer | NM_002529.2 |
| För användning med (applikation) | Functional Study, SDS-PAGE |
| Formulering | Liquid |
| Gen-ID (Entrez) | 4914 |
| Molekylvikt (g/mol) | 69.726kDa |
| Namn | NTRK1 (Human) Recombinant Protein |
| Beredningsmetod | Insect cell (Sf9) expression system |
| Reningsmetod | GST-Affinity Chromatography |
| Kvalitetskontrolltestning | 2 ug/lane SDS-PAGE Stained with Coomassie Blue |
| Kvantitet | 100 μg |
| Visa mer |
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.