Learn More
Abnova™ Human NUP210 Full-length ORF (AAH67089.1, 1 a.a. - 967 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023225-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex. [provided by RefSeq]
Sequence: MAARGRGLLLLTLSVLLAAGPSAAAAKLNIPKVLLPFTRATRVNFTLEASEGCYRWLSTRPEVASIEPLGLDEQQCSQKAVVQARLTQPARLTSIIFAEDITTGQVLRCDAIVDLIHDIQIVSTTRELYLEDSPLELKIQALDSEGNTFSTLAGLVFEWTIVKDSEADRFSDSHNALRILTFLESTYIPPSYISEMEKAAKQGDTILVSGMKTGSSKLKARIQEAVYKNVRPAEVRLLILENILLNPAYDVYLMVGTSIHYKVQKIRQGKITELSMPSDQYELQLQNSIPGPEGDPARPVAVLAQDTSMVTALQLGQSSLVLGHRSIRMQGASRLPNSTIYVVEPGYLGFTVHPGDRWVLETGRLYEITIEVFDKFSNKVYVSDNIRIETVLPAEFFEVLSSSQNGSYHRIRALKRGQTAIDAALTSVVDQDGGVHILQVPVWNQQEVEIHIPITLYPSILTFPWQPKTGAYQYTIRAHGGSGNFSWSSSSHLVATVTVKGVMTTGSDIGFSVIQAHDVQNPLHFGEMKVYVIEPHSMEFAPCQVEARVGQALELPLRISGLMPGGASEVVTLSDCSHFDLAVEVENQGVFQPLPGRLPPGSEHCSGIRVKAEAQGSTTLLVSYRHGHVHLSAKITIAAYLPLKAVDPSSVALVTLGSSKEMLFEGGPRPWILEPSKFFQNVTAEDTDSIGLALFAPHSSRNYQQHWILVTCQALGEQVIALSVGNKPSLTNPFPAVEPAVVKFVCAPPSRLTLAPVYTSPQLDMSCPLLQQNKQVVPVSSHRNPRLDLAAYDQEGRRFDNFSSLSIQWESTRPVLASIEPELPMQLVSQDDESGQKKLHGLQAILVHEASGTTAITATATGYQESHLSSARTKQPHDPLVPLSASIELILVEDVRVSPEEVTIYNHPGIQAELRIREGSGYFFLNTSTADVVKVAYQEARGVAMVSLGHRSPLLVFIPYLGCCVVNSpecifications
AAH67089.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
132.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ22389/GP210/KIAA0906/POM210 | |
NUP210 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23225 | |
NUP210 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAARGRGLLLLTLSVLLAAGPSAAAAKLNIPKVLLPFTRATRVNFTLEASEGCYRWLSTRPEVASIEPLGLDEQQCSQKAVVQARLTQPARLTSIIFAEDITTGQVLRCDAIVDLIHDIQIVSTTRELYLEDSPLELKIQALDSEGNTFSTLAGLVFEWTIVKDSEADRFSDSHNALRILTFLESTYIPPSYISEMEKAAKQGDTILVSGMKTGSSKLKARIQEAVYKNVRPAEVRLLILENILLNPAYDVYLMVGTSIHYKVQKIRQGKITELSMPSDQYELQLQNSIPGPEGDPARPVAVLAQDTSMVTALQLGQSSLVLGHRSIRMQGASRLPNSTIYVVEPGYLGFTVHPGDRWVLETGRLYEITIEVFDKFSNKVYVSDNIRIETVLPAEFFEVLSSSQNGSYHRIRALKRGQTAIDAALTSVVDQDGGVHILQVPVWNQQEVEIHIPITLYPSILTFPWQPKTGAYQYTIRAHGGSGNFSWSSSSHLVATVTVKGVMTTGSDIGFSVIQAHDVQNPLHFGEMKVYVIEPHSMEFAPCQVEARVGQALELPLRISGLMPGGASEVVTLSDCSHFDLAVEVENQGVFQPLPGRLPPGSEHCSGIRVKAEAQGSTTLLVSYRHGHVHLSAKITIAAYLPLKAVDPSSVALVTLGSSKEMLFEGGPRPWILEPSKFFQNVTAEDTDSIGLALFAPHSSRNYQQHWILVTCQALGEQVIALSVGNKPSLTNPFPAVEPAVVKFVCAPPSRLTLAPVYTSPQLDMSCPLLQQNKQVVPVSSHRNPRLDLAAYDQEGRRFDNFSSLSIQWESTRPVLASIEPELPMQLVSQDDESGQKKLHGLQAILVHEASGTTAITATATGYQESHLSSARTKQPHDPLVPLSASIELILVEDVRVSPEEVTIYNHPGIQAELRIREGSGYFFLNTSTADVVKVAYQEARGVAMVSLGHRSPLLVFIPYLGCCVVN | |
RUO | |
NUP210 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |