missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NUP210 Full-length ORF (AAH67089.1, 1 a.a. - 967 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH67089.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23225 |
Molecular Weight (g/mol) | 132.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16177072
|
Abnova™
H00023225-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16167072
|
Abnova™
H00023225-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex. [provided by RefSeq]
Sequence: MAARGRGLLLLTLSVLLAAGPSAAAAKLNIPKVLLPFTRATRVNFTLEASEGCYRWLSTRPEVASIEPLGLDEQQCSQKAVVQARLTQPARLTSIIFAEDITTGQVLRCDAIVDLIHDIQIVSTTRELYLEDSPLELKIQALDSEGNTFSTLAGLVFEWTIVKDSEADRFSDSHNALRILTFLESTYIPPSYISEMEKAAKQGDTILVSGMKTGSSKLKARIQEAVYKNVRPAEVRLLILENILLNPAYDVYLMVGTSIHYKVQKIRQGKITELSMPSDQYELQLQNSIPGPEGDPARPVAVLAQDTSMVTALQLGQSSLVLGHRSIRMQGASRLPNSTIYVVEPGYLGFTVHPGDRWVLETGRLYEITIEVFDKFSNKVYVSDNIRIETVLPAEFFEVLSSSQNGSYHRIRALKRGQTAIDAALTSVVDQDGGVHILQVPVWNQQEVEIHIPITLYPSILTFPWQPKTGAYQYTIRAHGGSGNFSWSSSSHLVATVTVKGVMTTGSDIGFSVIQAHDVQNPLHFGEMKVYVIEPHSMEFAPCQVEARVGQALELPLRISGLMPGGASEVVTLSDCSHFDLAVEVENQGVFQPLPGRLPPGSEHCSGIRVKAEAQGSTTLLVSYRHGHVHLSAKITIAAYLPLKAVDPSSVALVTLGSSKEMLFEGGPRPWILEPSKFFQNVTAEDTDSIGLALFAPHSSRNYQQHWILVTCQALGEQVIALSVGNKPSLTNPFPAVEPAVVKFVCAPPSRLTLAPVYTSPQLDMSCPLLQQNKQVVPVSSHRNPRLDLAAYDQEGRRFDNFSSLSIQWESTRPVLASIEPELPMQLVSQDDESGQKKLHGLQAILVHEASGTTAITATATGYQESHLSSARTKQPHDPLVPLSASIELILVEDVRVSPEEVTIYNHPGIQAELRIREGSGYFFLNTSTADVVKVAYQEARGVAMVSLGHRSPLLVFIPYLGCCVVNSpecifications
AAH67089.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
132.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAARGRGLLLLTLSVLLAAGPSAAAAKLNIPKVLLPFTRATRVNFTLEASEGCYRWLSTRPEVASIEPLGLDEQQCSQKAVVQARLTQPARLTSIIFAEDITTGQVLRCDAIVDLIHDIQIVSTTRELYLEDSPLELKIQALDSEGNTFSTLAGLVFEWTIVKDSEADRFSDSHNALRILTFLESTYIPPSYISEMEKAAKQGDTILVSGMKTGSSKLKARIQEAVYKNVRPAEVRLLILENILLNPAYDVYLMVGTSIHYKVQKIRQGKITELSMPSDQYELQLQNSIPGPEGDPARPVAVLAQDTSMVTALQLGQSSLVLGHRSIRMQGASRLPNSTIYVVEPGYLGFTVHPGDRWVLETGRLYEITIEVFDKFSNKVYVSDNIRIETVLPAEFFEVLSSSQNGSYHRIRALKRGQTAIDAALTSVVDQDGGVHILQVPVWNQQEVEIHIPITLYPSILTFPWQPKTGAYQYTIRAHGGSGNFSWSSSSHLVATVTVKGVMTTGSDIGFSVIQAHDVQNPLHFGEMKVYVIEPHSMEFAPCQVEARVGQALELPLRISGLMPGGASEVVTLSDCSHFDLAVEVENQGVFQPLPGRLPPGSEHCSGIRVKAEAQGSTTLLVSYRHGHVHLSAKITIAAYLPLKAVDPSSVALVTLGSSKEMLFEGGPRPWILEPSKFFQNVTAEDTDSIGLALFAPHSSRNYQQHWILVTCQALGEQVIALSVGNKPSLTNPFPAVEPAVVKFVCAPPSRLTLAPVYTSPQLDMSCPLLQQNKQVVPVSSHRNPRLDLAAYDQEGRRFDNFSSLSIQWESTRPVLASIEPELPMQLVSQDDESGQKKLHGLQAILVHEASGTTAITATATGYQESHLSSARTKQPHDPLVPLSASIELILVEDVRVSPEEVTIYNHPGIQAELRIREGSGYFFLNTSTADVVKVAYQEARGVAMVSLGHRSPLLVFIPYLGCCVVN | |
RUO | |
NUP210 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23225 | |
NUP210 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ22389/GP210/KIAA0906/POM210 | |
NUP210 | |
Recombinant | |
wheat germ expression system |