missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NUP43 Full-length ORF (NP_942590.1, 1 a.a. - 380 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_942590.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 348995 |
Molecular Weight (g/mol) | 68.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16170304
|
Abnova™
H00348995-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16180304
|
Abnova™
H00348995-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Bidirectional transport of macromolecules between the cytoplasm and nucleus occurs through nuclear pore complexes (NPCs) embedded in the nuclear envelope. NPCs are composed of subcomplexes, and NUP43 is part of one such subcomplex, Nup107-160 (Loiodice et al., 2004 [PubMed 15146057]).[supplied by OMIM]
Sequence: MEEIYAKFVSQKISKTRWRPLPPGSLQTAETFATGSWDNEENYISLWSIGDFGNLDSDGGFEGDHQLLCDIRHHGDVMDLQFFDQERIVAASSTGCVTVFLHHPNNQTLSVNQQWTTAHYHTGPGSPSYSSAPCTGVVCNNPEIVTVGEDGRINLFRADHKEAVRTIDNADSSTLHAVTFLRTPEILTVNSIGQLKIWDFRQQGNEPSQILSLTGDRVPLHCVDRHPNQQHVVATGGQDGMLSIWDVRQGTMPVSLLKAHEAEMWEVHFHPSNPEHLFTCSEDGSLWHWDASTDVPEKSSLFHQGGRSSTFLSHSISNQANVHQSVISSWLSTDPAKDRIEITSLLPSRSLSVNTLDVLGPCLVCGTDAEAIYVTRHLFSSpecifications
NP_942590.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
68.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEEIYAKFVSQKISKTRWRPLPPGSLQTAETFATGSWDNEENYISLWSIGDFGNLDSDGGFEGDHQLLCDIRHHGDVMDLQFFDQERIVAASSTGCVTVFLHHPNNQTLSVNQQWTTAHYHTGPGSPSYSSAPCTGVVCNNPEIVTVGEDGRINLFRADHKEAVRTIDNADSSTLHAVTFLRTPEILTVNSIGQLKIWDFRQQGNEPSQILSLTGDRVPLHCVDRHPNQQHVVATGGQDGMLSIWDVRQGTMPVSLLKAHEAEMWEVHFHPSNPEHLFTCSEDGSLWHWDASTDVPEKSSLFHQGGRSSTFLSHSISNQANVHQSVISSWLSTDPAKDRIEITSLLPSRSLSVNTLDVLGPCLVCGTDAEAIYVTRHLFS | |
RUO | |
NUP43 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
348995 | |
NUP43 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ13287/bA350J20.1/p42 | |
NUP43 | |
Recombinant | |
wheat germ expression system |