missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NXF3 Full-length ORF (AAH31616, 1 a.a. - 531 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056000-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16182013

  • 4515.00 SEK / 10µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is one member of a family of nuclear RNA export factor genes. Common domain features of this family are a noncanonical RNP-type RNA-binding domain (RBD), 4 leucine-rich repeats (LRRs), a nuclear transport factor 2 (NTF2)-like domain that allows heterodimerization with NTF2-related export protein-1 (NXT1), and a ubiquitin-associated domain that mediates interactions with nucleoporins. The LRRs and NTF2-like domains are required for export activity. Alternative splicing seems to be a common mechanism in this gene family. The encoded protein of this gene has shortened LRR and ubiquitin-associated domains and its RDB is unable to bind RNA. It is located in the nucleoplasm but is not associated with either the nuclear envelope or the nucleolus. [provided by RefSeq]

Sequence: MSLPSGHTTGHTDQVVQRRARCWDIYQRRFSSRSEPVNPGMHSSSHQQQDGDAAMHGAHMDSPVRYTPYTISPYNRKGSFRKQDQTHVNMEREQKPPERRMEGNMPDGTLGSWFKITVPFGIKYNEKWLLNLIQNECSVPFVPVEFHYENMHASFFVENASIAYALKNVSGKIWDEDNEKISIFVNPAGIPHFVHRELKSEKVEQIKLAMNQQCDVSQEALDIQRLPFYPDMVNRDTKMASNPRKCMAASLDVHEENIPTVMSAGEMDKWKGIEPGEKCADRSPVCTTFSDTSSNINSILELFPKLLCLDGQQSPRATLCGTEAHKRLPTCKGSFFGSEMLKNLVLQFLQQYYLIYDSGDRQGLLSAYHDEACFSLSIPFNPEDSAPSSFCKFFKDSRNIKILKDPYLRGELLKHTKLDIVDSLSALPKTQHDLSSFLVDMWYQTEWMLCFSVNGVFKEVEGQSQGSVLAFTRTFIATPGSSSSLCIVNDKLFVRDTSHQGTQSALFTLVPTAFSSSMPAFSQEQQKMLPS
Specifications

Specifications

AAH31616
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.15kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NXF3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
56000
NXF3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSLPSGHTTGHTDQVVQRRARCWDIYQRRFSSRSEPVNPGMHSSSHQQQDGDAAMHGAHMDSPVRYTPYTISPYNRKGSFRKQDQTHVNMEREQKPPERRMEGNMPDGTLGSWFKITVPFGIKYNEKWLLNLIQNECSVPFVPVEFHYENMHASFFVENASIAYALKNVSGKIWDEDNEKISIFVNPAGIPHFVHRELKSEKVEQIKLAMNQQCDVSQEALDIQRLPFYPDMVNRDTKMASNPRKCMAASLDVHEENIPTVMSAGEMDKWKGIEPGEKCADRSPVCTTFSDTSSNINSILELFPKLLCLDGQQSPRATLCGTEAHKRLPTCKGSFFGSEMLKNLVLQFLQQYYLIYDSGDRQGLLSAYHDEACFSLSIPFNPEDSAPSSFCKFFKDSRNIKILKDPYLRGELLKHTKLDIVDSLSALPKTQHDLSSFLVDMWYQTEWMLCFSVNGVFKEVEGQSQGSVLAFTRTFIATPGSSSSLCIVNDKLFVRDTSHQGTQSALFTLVPTAFSSSMPAFSQEQQKMLPS
RUO
NXF3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers