Learn More
Abnova™ Human OATL1 Partial ORF (NP_002527.1, 302 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004943-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein with a TBC domain and may function as a Rab GTPase activating protein. This gene was previously known as ornithine aminotransferase-like 1, but has no similarity to ornithine aminotransferase. [provided by RefSeq]
Sequence: YAGPEDGPHLRALHDLLTTYAVTHPQVSYCQGMSDLASPILAVMDHEGHAFVCFCGIMKRLAANFHPDGRAMATKFAHLKLLLRHADPDFYQYLQEAGADDLFFCYRWLSpecifications
NP_002527.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YAGPEDGPHLRALHDLLTTYAVTHPQVSYCQGMSDLASPILAVMDHEGHAFVCFCGIMKRLAANFHPDGRAMATKFAHLKLLLRHADPDFYQYLQEAGADDLFFCYRWL | |
RUO | |
TBC1D25 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4943 | |
OATL1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MG81/MGC126866/MGC126868/MGC149731/MGC149732/OATL1 | |
TBC1D25 | |
Recombinant | |
wheat germ expression system |