missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OCA2 Partial ORF (AAH12097, 201 a.a. - 300 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH12097 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4948 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16178551
|
Abnova™
H00004948-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16168551
|
Abnova™
H00004948-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes the human homologue of the mouse p (pink-eyed dilution) gene. The encoded protein is believed to be an integral membrane protein involved in small molecule transport, specifically tyrosine - a precursor of melanin. Mutations in this gene result in type 2 oculocutaneous albinism. [provided by RefSeq]
Sequence: GKLWQLLALSPLENYSVNLSSHVDSTLLQVDLAGALVASGPSRPGREEHIVVELTQADALGSRWRRPQQVTHNWTVYLNPRRSEHSVMSRTFEVLTRETVSpecifications
AAH12097 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BEY/BEY1/BEY2/BOCA/D15S12/EYCL/EYCL2/EYCL3/HCL3/P/PED/SHEP1 | |
OCA2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4948 | |
OCA2 (Human) Recombinant Protein (Q01) | |
GKLWQLLALSPLENYSVNLSSHVDSTLLQVDLAGALVASGPSRPGREEHIVVELTQADALGSRWRRPQQVTHNWTVYLNPRRSEHSVMSRTFEVLTRETV | |
RUO | |
OCA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |