missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ODF4 Full-length ORF (NP_694552.1, 1 a.a. - 257 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00146852-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16118443

  • 6845.00 SEK / 25µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that is localized in the outer dense fibers of the tails of mature sperm. This protein is thought to have some important role in the sperm tail. [provided by RefSeq]

Sequence: MDAEYSGNEFPRSEGERDQHQRPGKERKSGEAGWGTGELGQDGRLLSSTLSLSSNRSLGQRQNSPLPFQWRITHSFRWMAQVLASELSLVAFILLLVVAFSKKWLDLSRSLFYQRWPVDVSNRIHTSAHVMSMGLLHFYKSRSCSDLENGKVTFIFSTLMLFPINIWIFELERNVSIPIGWSYFIGWLVLILYFTCAILCYFNHKSFWSLILSHPSGAVSCSSSFGSVEESPRAQTITDTPITQEGVLDPEQKDTHV
Specifications

Specifications

NP_694552.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
55.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC138215/MGC138219/OPPO1
ODF4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
146852
ODF4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDAEYSGNEFPRSEGERDQHQRPGKERKSGEAGWGTGELGQDGRLLSSTLSLSSNRSLGQRQNSPLPFQWRITHSFRWMAQVLASELSLVAFILLLVVAFSKKWLDLSRSLFYQRWPVDVSNRIHTSAHVMSMGLLHFYKSRSCSDLENGKVTFIFSTLMLFPINIWIFELERNVSIPIGWSYFIGWLVLILYFTCAILCYFNHKSFWSLILSHPSGAVSCSSSFGSVEESPRAQTITDTPITQEGVLDPEQKDTHV
RUO
ODF4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers