missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OGN Partial ORF (NP_148935.1, 200 a.a. - 298 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_148935.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4969 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16132195
|
Abnova™
H00004969-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16142195
|
Abnova™
H00004969-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein which induces ectopic bone formation in conjunction with transforming growth factor beta. This protein is a small proteoglycan which contains tandem leucine-rich repeats (LRR). Alternatively spliced transcript variants that encode the same protein have been found for this gene. [provided by RefSeq]
Sequence: KSRGIKANAFKKLNNLTFLYLDHNALESVPLNLPESLRVIHLQFNNIASITDDTFCKANDTSYIRDRIEEIRLEGNPIVLGKHPNSFICLKRLPIGSYFSpecifications
NP_148935.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586P2421/OG/OIF/SLRR3A | |
OGN | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4969 | |
OGN (Human) Recombinant Protein (Q01) | |
KSRGIKANAFKKLNNLTFLYLDHNALESVPLNLPESLRVIHLQFNNIASITDDTFCKANDTSYIRDRIEEIRLEGNPIVLGKHPNSFICLKRLPIGSYF | |
RUO | |
OGN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |