missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OMP Full-length ORF (NP_006180.1, 1 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_006180.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4975 |
Molecular Weight (g/mol) | 45.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16108581
|
Abnova™
H00004975-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16198571
|
Abnova™
H00004975-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Olfactory marker protein is uniquely associated with the mature olfactory receptor neurons in many vertebrate species from fish to man. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Results of the mouse knockout studies show that OMP-null mice are compromised in their ability to respond to odor stimuli, and that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. [provided by RefSeq]
Sequence: MAEDRPQQPQLDMPLVLDQGLTRQMRLRVESLKQRGEKRQDGEKLLQPAESVYRLNFTQQQRLQFERWNVVLDKPGKVTITGTSQNWTPDLTNLMTRQLLDPTAIFWRKEDSDAIDWNEADALEFGERLSDLAKIRKVMYFLVTFGEGVEPANLKASVVFNQLSpecifications
NP_006180.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAEDRPQQPQLDMPLVLDQGLTRQMRLRVESLKQRGEKRQDGEKLLQPAESVYRLNFTQQQRLQFERWNVVLDKPGKVTITGTSQNWTPDLTNLMTRQLLDPTAIFWRKEDSDAIDWNEADALEFGERLSDLAKIRKVMYFLVTFGEGVEPANLKASVVFNQL | |
RUO | |
OMP | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4975 | |
OMP (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
OMP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |