Learn More
Abnova™ Human OPCML Full-length ORF (AAH74742.1, 1 a.a. - 344 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004978-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the IgLON subfamily in the immunoglobulin protein superfamily. The encoded protein is localized in the plasma membrane and may have an accessory role in opioid receptor function. This gene has an ortholog in rat and bovine. The opioid binding-cell adhesion molecule encoded by the rat gene binds opioid alkaloids in the presence of acidic lipids, exhibits selectivity for mu ligands and acts as a GPI-anchored protein. Since the encoded protein is highly conserved in species during evolution, it may have a fundamental role in mammalian systems. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MGVCGYLFLPWKCLVVVSLRLLFLVPTGVPVRSGDATFPKAMDNVTVRQGESATLRCTIDDRVTRVAWLNRSTILYAGNDKWSIDPRVIILVNTPTQYSIMIQNVDVYDEGPYTCSVQTDNHPKTSRVHLIVQVPPQIMNISSDITVNEGSSVTLLCLAIGRSEPTVTWRHLSVKGQGFVSEDEYLEISDIKRDQSGEYECSALNDVAAPDVRKVKITVNYPPYISKAKNTGVSVGQKGILSCEASAVPMAEFQWFKEETRLATGLDGMRIENKGRMSTLTFFNVSEKDYGNYTCVATNKLGNTNASITLYGPGAVIDGVNSASRALACLWLSGTLLAHFFIKFSpecifications
AAH74742.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IGLON1/OBCAM/OPCM | |
OPCML | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4978 | |
OPCML (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGVCGYLFLPWKCLVVVSLRLLFLVPTGVPVRSGDATFPKAMDNVTVRQGESATLRCTIDDRVTRVAWLNRSTILYAGNDKWSIDPRVIILVNTPTQYSIMIQNVDVYDEGPYTCSVQTDNHPKTSRVHLIVQVPPQIMNISSDITVNEGSSVTLLCLAIGRSEPTVTWRHLSVKGQGFVSEDEYLEISDIKRDQSGEYECSALNDVAAPDVRKVKITVNYPPYISKAKNTGVSVGQKGILSCEASAVPMAEFQWFKEETRLATGLDGMRIENKGRMSTLTFFNVSEKDYGNYTCVATNKLGNTNASITLYGPGAVIDGVNSASRALACLWLSGTLLAHFFIKF | |
RUO | |
OPCML | |
Wheat Germ (in vitro) | |
GST | |
Liquid |