missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OPHN1 Full-length ORF (NP_002538.1, 1 a.a. - 802 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004983-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16118591

  • 4515.00 SEK / 10µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a Rho-GTPase-activating protein that promotes GTP hydrolysis of Rho subfamily members. Rho proteins are important mediators of intracellular signal transduction, which affects cell migration and cell morphogenesis. Mutations in this gene are responsible for OPHN1-related X-linked mental retardation with cerebellar hypoplasia and distinctive facial dysmorhphism. [provided by RefSeq]

Sequence: MGHPPLEFSDCYLDSPDFRERLKCYEQELERTNKFIKDVIKDGNALISAMRNYSSAVQKFSQTLQSFQFDFIGDTLTDDEINIAESFKEFAELLNEVENERMMMVHNASDLLIKPLENFRKEQIGFTKERKKKFEKDGERFYSLLDRHLHLSSKKKESQLQEADLQVDKERHNFFESSLDYVYQIQEVQESKKFNIVEPVLAFLHSLFISNSLTVELTQDFLPYKQQLQLSLQNTRNHFSSTREEMEELKKRMKEAPQTCKLPGQPTIEGYLYTQEKWALGISWVKYYCQYEKETKTLTMTPMEQKPGAKQGPLDLTLKYCVRRKTESIDKRFCFDIETNERPGTITLQALSEANRRLWMEAMDGKEPIYHSPITKQQEMELNEVGFKFVRKCINIIETKGIKTEGLYRTVGSNIQVQKLLNAFFDPKCPGDVDFHNSDWDIKTITSSLKFYLRNLSEPVMTYRLHKELVSAAKSDNLDYRLGAIHSLVYKLPEKNREMLELLIRHLVNVCEHSKENLMTPSNMGVIFGPTLMRAQEDTVAAMMNIKFQNIVVEILIEHFGKIYLGPPEESAAPPVPPPRVTARRHKPITISKRLLRERTVFYTSSLDESEDEIQHQTPNGTITSSIEPPKPPQHPKLPIQRSGETDPGRKSPSRPILDGKLEPCPEVDVGKLVSRLQDGGTKITPKATNGPMPGSGPTKTPSFHIKRPAPRPLAHHKEGDADSFSKVRPPGEKPTIIRPPVRPPDPPCRAATPQKPEPKPDIVAGNAGEITSSVVASRTRFFETASRKTGSSQGRLPGDES
Specifications

Specifications

NP_002538.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MRX60/OPN1
OPHN1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
4983
OPHN1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGHPPLEFSDCYLDSPDFRERLKCYEQELERTNKFIKDVIKDGNALISAMRNYSSAVQKFSQTLQSFQFDFIGDTLTDDEINIAESFKEFAELLNEVENERMMMVHNASDLLIKPLENFRKEQIGFTKERKKKFEKDGERFYSLLDRHLHLSSKKKESQLQEADLQVDKERHNFFESSLDYVYQIQEVQESKKFNIVEPVLAFLHSLFISNSLTVELTQDFLPYKQQLQLSLQNTRNHFSSTREEMEELKKRMKEAPQTCKLPGQPTIEGYLYTQEKWALGISWVKYYCQYEKETKTLTMTPMEQKPGAKQGPLDLTLKYCVRRKTESIDKRFCFDIETNERPGTITLQALSEANRRLWMEAMDGKEPIYHSPITKQQEMELNEVGFKFVRKCINIIETKGIKTEGLYRTVGSNIQVQKLLNAFFDPKCPGDVDFHNSDWDIKTITSSLKFYLRNLSEPVMTYRLHKELVSAAKSDNLDYRLGAIHSLVYKLPEKNREMLELLIRHLVNVCEHSKENLMTPSNMGVIFGPTLMRAQEDTVAAMMNIKFQNIVVEILIEHFGKIYLGPPEESAAPPVPPPRVTARRHKPITISKRLLRERTVFYTSSLDESEDEIQHQTPNGTITSSIEPPKPPQHPKLPIQRSGETDPGRKSPSRPILDGKLEPCPEVDVGKLVSRLQDGGTKITPKATNGPMPGSGPTKTPSFHIKRPAPRPLAHHKEGDADSFSKVRPPGEKPTIIRPPVRPPDPPCRAATPQKPEPKPDIVAGNAGEITSSVVASRTRFFETASRKTGSSQGRLPGDES
RUO
OPHN1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers