missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OR2J1 Full-length ORF (Q9GZK6, 1 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00442185-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120744

  • 4055.00 SEK / 10µg
Estimated Shipment: 14-07-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]

Sequence: MLMKKNASFEDFFILLGFSNWPHLEVVLFVVILIFYLITLIGNLFIIILSYLDSHLHTPMYFFLSNLSFLDLCYTTSSIPQLLVNLWGPEKTISYAGCTVQLYFVLALGTAECVLLVVMSYDRYAAVCRPLHYTVLMHPRFCRLLAAASWVSGFTTSALHSSFTFWIPLCRHRLVDHFFCEVPALLRLSCVDTQANELTLMVMSSIFVLIPLILILTSYGAIARAVLSMQSTTGLQKVLRTCGAHLMVVSLFFIPVMCMYLQPPSENSQDQGKFIALFYTVVTPSLNPLIYTFRNKDVRGAVKRLMGWEWGM
Specifications

Specifications

Q9GZK6
Liquid
442185
OR2J1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLMKKNASFEDFFILLGFSNWPHLEVVLFVVILIFYLITLIGNLFIIILSYLDSHLHTPMYFFLSNLSFLDLCYTTSSIPQLLVNLWGPEKTISYAGCTVQLYFVLALGTAECVLLVVMSYDRYAAVCRPLHYTVLMHPRFCRLLAAASWVSGFTTSALHSSFTFWIPLCRHRLVDHFFCEVPALLRLSCVDTQANELTLMVMSSIFVLIPLILILTSYGAIARAVLSMQSTTGLQKVLRTCGAHLMVVSLFFIPVMCMYLQPPSENSQDQGKFIALFYTVVTPSLNPLIYTFRNKDVRGAVKRLMGWEWGM
RUO
OR2J1
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
6M1-4P/OR2J1P/OR6-15/OR6-5/dJ80I19.2/hs6M1-4
OR2J1
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers