Learn More
Abnova™ Human OR6N1 Full-length ORF (NP_001005185.1, 1 a.a. - 312 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_001005185.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 128372 |
Molecular Weight (g/mol) | 61.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137813
|
Abnova™
H00128372-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16127813
|
Abnova™
H00128372-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq]
Sequence: MDTGNWSQVAEFIILGFPHLQGVQIYLFLLLLLIYLMTVLGNLLIFLVVCLDSRLHTPMYHFVSILSFSELGYTAATIPKMLANLLSEKKTISFSGCLLQIYFFHSLGATECYLLTAMAYDRYLAICRPLHYPTLMTPTLCAEIAIGCWLGGLAGPVVEISLISRLPFCGPNRIQHVFCDFPPVLSLACTDTSINVLVDFVINSCKILATFLLILCSYVQIICTVLRIPSAAGKRKAISTCASHFTVVLIFYGSILSMYVQLKKSYSLDYDQALAVVYSVLTPFLNPFIYSLRNKEIKEAVRRQLKRIGILASpecifications
NP_001005185.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
61.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDTGNWSQVAEFIILGFPHLQGVQIYLFLLLLLIYLMTVLGNLLIFLVVCLDSRLHTPMYHFVSILSFSELGYTAATIPKMLANLLSEKKTISFSGCLLQIYFFHSLGATECYLLTAMAYDRYLAICRPLHYPTLMTPTLCAEIAIGCWLGGLAGPVVEISLISRLPFCGPNRIQHVFCDFPPVLSLACTDTSINVLVDFVINSCKILATFLLILCSYVQIICTVLRIPSAAGKRKAISTCASHFTVVLIFYGSILSMYVQLKKSYSLDYDQALAVVYSVLTPFLNPFIYSLRNKEIKEAVRRQLKRIGILA | |
RUO | |
OR6N1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
128372 | |
OR6N1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
OR1-22 | |
OR6N1 | |
Recombinant | |
wheat germ expression system |