missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OSBPL1A Full-length ORF (AAH41563.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00114876-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16177133

  • 5030.41 SEK / 25µg

Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Transcript variants derived from alternative promoter usage and/or alternative splicing exist; they encode different isoforms. [provided by RefSeq]

Sequence: MNTEAEQQLLHHARNGNAEEVRQLLETMARNEVIADINCKGRSKSNLGWTPLHLACYFGHRQVVQDLLKAGAEVNVLNDMRDTPLHRAAFTGRKICSQGS
Specifications

Specifications

AAH41563.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10217/ORP-1/ORP1/OSBPL1B
OSBPL1A
Yes
wheat germ expression system
Liquid
114876
OSBPL1A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNTEAEQQLLHHARNGNAEEVRQLLETMARNEVIADINCKGRSKSNLGWTPLHLACYFGHRQVVQDLLKAGAEVNVLNDMRDTPLHRAAFTGRKICSQGS
RUO
OSBPL1A
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates