missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OSBPL1A Full-length ORF (AAH41563.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH41563.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 114876 |
Molecular Weight (g/mol) | 37.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16167133
|
Abnova™
H00114876-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16177133
|
Abnova™
H00114876-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Transcript variants derived from alternative promoter usage and/or alternative splicing exist; they encode different isoforms. [provided by RefSeq]
Sequence: MNTEAEQQLLHHARNGNAEEVRQLLETMARNEVIADINCKGRSKSNLGWTPLHLACYFGHRQVVQDLLKAGAEVNVLNDMRDTPLHRAAFTGRKICSQGSSpecifications
AAH41563.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNTEAEQQLLHHARNGNAEEVRQLLETMARNEVIADINCKGRSKSNLGWTPLHLACYFGHRQVVQDLLKAGAEVNVLNDMRDTPLHRAAFTGRKICSQGS | |
RUO | |
OSBPL1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
114876 | |
OSBPL1A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10217/ORP-1/ORP1/OSBPL1B | |
OSBPL1A | |
Recombinant | |
wheat germ expression system |