missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human OSBPL1A Partial ORF (NP_542164.2, 268 a.a. - 340 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_542164.2 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 114876 |
Molecular Weight (g/mol) | 33.77kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16101437
|
Abnova™
H00114876-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16191427
|
Abnova™
H00114876-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Transcript variants derived from alternative promoter usage and/or alternative splicing exist; they encode different isoforms. [provided by RefSeq]
Sequence: KQPDAVHNIYRQGCKHLTQAVCTVKSTDSCLFFIKCFDDTIHGFRVPKNSLQQSREDWLEAIEEHSAYSTHYCSpecifications
NP_542164.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.77kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10217/ORP-1/ORP1/OSBPL1B | |
OSBPL1A | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
114876 | |
OSBPL1A (Human) Recombinant Protein (Q01) | |
KQPDAVHNIYRQGCKHLTQAVCTVKSTDSCLFFIKCFDDTIHGFRVPKNSLQQSREDWLEAIEEHSAYSTHYC | |
RUO | |
OSBPL1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |