missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OSBPL2 Partial ORF (NP_653081.1, 294 a.a. - 393 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009885-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121416

  • 4515.00 SEK / 10µg
Estimated Shipment: 02-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. This encoded protein contains only the sterol-binding domain. In vitro studies have shown that the encoded protein can bind strongly to phosphatic acid and weakly to phosphatidylinositol 3-phosphate, but cannot bind to 25-hydroxycholesterol. The protein associates with the Golgi apparatus. Transcript variants encoding different isoforms have been described. [provided by RefSeq]

Sequence: KLFMIYGKWTECLWGIDPVSYESFKKQERRGDHLRKAKLDEDSGKADSDVADDVPVAQETVQVIPGSKLLWRINTRPPNSAQMYNFTSFTVSLNELETGM
Specifications

Specifications

NP_653081.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KLFMIYGKWTECLWGIDPVSYESFKKQERRGDHLRKAKLDEDSGKADSDVADDVPVAQETVQVIPGSKLLWRINTRPPNSAQMYNFTSFTVSLNELETGM
RUO
OSBPL2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9885
OSBPL2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ20223/KIAA0772/MGC4307/MGC8342/ORP-2/ORP2
OSBPL2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers