Learn More
Abnova™ Human OSCAR Partial ORF (NP_996553.1, 175 a.a. - 254 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00126014-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Osteoclasts are multinucleated cells that resorb bone and are essential for bone homeostasis. This gene encodes an osteoclast-associated receptor (OSCAR), which is a member of the leukocyte receptor complex (LRC) protein family that plays critical roles in the regulation of both innate and adaptive immune responses. Different from the other LRC members, OSCAR expression is detected specifically in preosteoclasts or mature osteoclasts. OSCAR may be an important bone-specific regulator of osteoclast differentiation. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: TLLGARAPGTYSCYYHTPSAPYVLSQRSEVLVISWEGEGPEARPASSAPGMQAPGPPPSDPGAQAPSLSSFRPRGLVLQPSpecifications
NP_996553.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.54kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TLLGARAPGTYSCYYHTPSAPYVLSQRSEVLVISWEGEGPEARPASSAPGMQAPGPPPSDPGAQAPSLSSFRPRGLVLQP | |
RUO | |
OSCAR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
126014 | |
OSCAR (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC33613/PIGR3 | |
OSCAR | |
Recombinant | |
wheat germ expression system |