missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human Osteoprotegerin/TNFRSF11B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8052-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifikationer
| Osteoprotegerin/TNFRSF11B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| OCIF, OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TNFRSF11B, TR1, tumor necrosis factor receptor superfamily, member 11b | |
| Mouse myeloma cell line, NS0-derived human Osteoprotegerin/TNFRSF11B, Glu22-Leu401, Accession # AAB53709.1 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 155321 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| AAB53709.1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 4982 | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering