Learn More
Abnova™ Human PADI3 Full-length ORF (NP_057317.2, 1 a.a. - 664 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051702-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type III enzyme modulates hair structural proteins, such as filaggrin in the hair follicle and trichohyalin in the inner root sheath, during hair follicle formation. Together with the type I enzyme, this enzyme may also play a role in terminal differentiation of the epidermis. This gene exists in a cluster with four other paralogous genes. [provided by RefSeq]
Sequence: MSLQRIVRVSLEHPTSAVCVAGVETLVDIYGSVPEGTEMFEVYGTPGVDIYISPNMERGRERADTRRWRFDATLEIIVVMNSPSNDLNDSHVQISYHSSHEPLPLAYAVLYLTCVDISLDCDLNCEGRQDRNFVDKRQWVWGPSGYGGILLVNCDRDDPSCDVQDNCDQHVHCLQDLEDMSVMVLRTQGPAALFDDHKLVLHTSSYDAKRAQVFHICGPEDVCEAYRHVLGQDKVSYEVPRLHGDEERFFVEGLSFPDAGFTGLISFHVTLLDDSNEDFSASPIFTDTVVFRVAPWIMTPSTLPPLEVYVCRVRNNTCFVDAVAELARKAGCKLTICPQAENRNDRWIQDEMELGYVQAPHKTLPVVFDSPRNGELQDFPYKRILGPDFGYVTREPRDRSVSGLDSFGNLEVSPPVVANGKEYPLGRILIGGNLPGSSGRRVTQVVRDFLHAQKVQPPVELFVDWLAVGHVDEFLSFVPAPDGKGFRMLLASPGACFKLFQEKQKCGHGRALLFQGVVDDEQVKTISINQVLSNKDLINYNKFVQSCIDWNREVLKRELGLAECDIIDIPQLFKTERKKATAFFPDLVNMLVLGKHLGIPKPFGPIINGCCCLEEKVRSLLEPLGLHCTFIDDFTPYHMLHGEVHCGTNVCRKPFSFKWWNMVPSpecifications
NP_057317.2 | |
Liquid | |
51702 | |
PADI3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSLQRIVRVSLEHPTSAVCVAGVETLVDIYGSVPEGTEMFEVYGTPGVDIYISPNMERGRERADTRRWRFDATLEIIVVMNSPSNDLNDSHVQISYHSSHEPLPLAYAVLYLTCVDISLDCDLNCEGRQDRNFVDKRQWVWGPSGYGGILLVNCDRDDPSCDVQDNCDQHVHCLQDLEDMSVMVLRTQGPAALFDDHKLVLHTSSYDAKRAQVFHICGPEDVCEAYRHVLGQDKVSYEVPRLHGDEERFFVEGLSFPDAGFTGLISFHVTLLDDSNEDFSASPIFTDTVVFRVAPWIMTPSTLPPLEVYVCRVRNNTCFVDAVAELARKAGCKLTICPQAENRNDRWIQDEMELGYVQAPHKTLPVVFDSPRNGELQDFPYKRILGPDFGYVTREPRDRSVSGLDSFGNLEVSPPVVANGKEYPLGRILIGGNLPGSSGRRVTQVVRDFLHAQKVQPPVELFVDWLAVGHVDEFLSFVPAPDGKGFRMLLASPGACFKLFQEKQKCGHGRALLFQGVVDDEQVKTISINQVLSNKDLINYNKFVQSCIDWNREVLKRELGLAECDIIDIPQLFKTERKKATAFFPDLVNMLVLGKHLGIPKPFGPIINGCCCLEEKVRSLLEPLGLHCTFIDDFTPYHMLHGEVHCGTNVCRKPFSFKWWNMVP | |
RUO | |
PADI3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
101.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126307/MGC126308/PAD3/PDI3 | |
PADI3 | |
Yes | |
wheat germ expression system |