Learn More
Abnova™ Human PAFAH2 Partial ORF (NP_000428, 293 a.a. - 392 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_000428 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5051 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16168671
|
Abnova™
H00005051-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16178671
|
Abnova™
H00005051-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes platelet-activating factor acetylhydrolase isoform 2, a single-subunit intracellular enzyme that catalyzes the removal of the acetyl group at the SN-2 position of platelet-activating factor (identified as 1-O-alkyl-2-acetyl-sn-glyceryl-3-phosphorylcholine). However, this lipase exhibits a broader substrate specificity than simply platelet activating factor. Two other isoforms of intracellular platelet-activating factor acetylhydrolase exist, and both are multi-subunit enzymes. Additionally, there is a single-subunit serum isoform of this enzyme. [provided by RefSeq]
Sequence: LMKKICAQHEQSRIITVLGSVHRSQTDFAFVTGNLIGKFFSTETRGSLDPYEGQEVMVRAMLAFLQKHLDLKEDYNQWNNLIEGIGPSLTPGAPHHLSSLSpecifications
NP_000428 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ26025/HSD-PLA2 | |
PAFAH2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
5051 | |
PAFAH2 (Human) Recombinant Protein (Q01) | |
LMKKICAQHEQSRIITVLGSVHRSQTDFAFVTGNLIGKFFSTETRGSLDPYEGQEVMVRAMLAFLQKHLDLKEDYNQWNNLIEGIGPSLTPGAPHHLSSL | |
RUO | |
PAFAH2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |